Human CD155 (from CHO cells)
Supplier: 0000027161
ANC-555-030
102979-340EA 750.46 USD
102979-340
Human CD155 (from CHO cells)
Proteins and Peptides
Human CD155 (Polio Virus Receptor, PVR, Necl-5) is a 70kd type I Ig superfamily molecule. It is involved in formation of intracellular junctions between epithelial cells. Its ligands include CD226 (DNAM-1), and CD96 (TACTILE). CD155 expression by tumor has been shown to be upregulated by nitric oxide. A soluble version of CD155 has been shown to exist. High CD155 expression has recently been exploited to use engineered poliovirus to treat glioblastoma.