Anti-CD155 Monoclonal Antibody (Biotin) [clone: ANC6A3]

Supplier: Adipogen
ANC-350-030
102979-320EA 483.23 USD
102979-320
Anti-CD155 Monoclonal Antibody (Biotin) [clone: ANC6A3]
Antibodies
Human CD155 (Polio Virus Receptor, PVR, Necl-5) is a 70 kd type I Ig superfamily molecule. It is involved in formation of intracellular junctions between epithelial cells. Its ligands include CD226 (DNAM-1), and CD96 (TACTILE). CD155 expression by tumor has been shown to be upregulated by nitric oxide. A soluble version of CD155 has been shown to exist. High CD155 expression has recently been exploited to use engineered poliovirus to treat glioblastoma.

Type: Primary
Antigen: CD155
Clonality: Monoclonal
Clone: ANC6A3
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR