Anti-ACR Monoclonal Antibody [clone: 5F6]

Supplier: Adipogen
JAI-MAR-020N
102979-296EA 743.54 USD
102979-296
Anti-ACR Monoclonal Antibody [clone: 5F6]
Antibodies
Acrolein (ACR) is a representative carcinogenic aldehyde found ubiquitously in the environment and formed endogenously through oxidation reactions, such as lipid peroxidation and myeloperoxidasecatalyzed amino acid oxidation. ACR is highly reactive aldehyde and reacts with lysine residue in protein. The reaction with ACR and lysine residue leads to the formation of numerous numbers of adducts, such as formyl-dehydropiperidino-lysine (FDP-lysine) type derivative.

Type: Primary
Antigen: ACR
Clonality: Monoclonal
Clone: 5F6
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: All, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR