Anti-IgG Goat Polyclonal Antibody (R-PE)

Supplier: Adipogen
AG-29B-0004R-C050
102978-662EA 221.54 USD
102978-662
Anti-IgG Goat Polyclonal Antibody (R-PE)
Antibodies
The anti-IgG (mouse), pAb (R-PE) is cross-absorbed against mouse IgM and IgA, pooled human sera and purified human paraproteins and affinity-purified on pooled mouse IgG covalently linked to agarose.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: R-PE
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR