Anti-IgG Goat Polyclonal Antibody (Horseradish peroxidase)

Supplier: Adipogen
AG-29B-0001E-R050
102978-654EA 221.54 USD
102978-654
Anti-IgG Goat Polyclonal Antibody (Horseradish peroxidase)
Antibodies
The anti-IgG (rat), pAb (HRP) is cross-adsorbed against pooled mouse sera and pooled mouse plasmacytoma/hybridoma proteins and affinity-purified on pooled rat IgG covalently linked to agarose.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Horseradish peroxidase
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR