Anti-CASP11 Monoclonal Antibody [clone: 4E11]

Supplier: Adipogen
AG-20T-0140-C100
102978-636EA 443.08 USD
102978-636
Anti-CASP11 Monoclonal Antibody [clone: 4E11]
Antibodies
Caspase-11 not only activates caspase-1, that is required for the maturation of proinflammatory cytokines such as interleukin-1 (IL-1) and interleukin-18 (IL-18), but also activates caspase-3, leading to cellular apoptosis under pathological conditions. In most cells, caspase-11 is only expressed upon induction with pro-inflammatory stimuli. Caspase-11 is an essential mediator of endotoxic shock and caspase-11-deficient mice are resistant to endotoxic shock. LPS-induced caspase-11 regulates lymphocyte apoptosis by activating both caspase-3 and caspase-7.

Type: Primary
Antigen: CASP11
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Rat IgG1kappa
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR