Anti-CASP8 Monoclonal Antibody [clone: 3B10]

Supplier: Adipogen
AG-20T-0138-C100
102978-632EA 401.54 USD
102978-632
Anti-CASP8 Monoclonal Antibody [clone: 3B10]
Antibodies
Procaspase-8 belongs to the family of caspases. Binding of FasL to Fas leads to formation of a receptor complex at the cellular membrane, which was named DISC. The DISC consists of oligomerized receptors, the DD-containing adaptor molecule FADD, procaspase-8, procaspase-10 and c-FLIP. The DISC structure provides a platform for the oligomerization of procaspase-8 that allows two procaspase-8 homodimers to be in the close proximity leading to the initial activation of procaspase-8. At the first cleavage step, the N-terminal p43/p41 and the C-terminal p30 cleavage products are generated. Importantly, these cleavage products already possess catalytic activity. At the second cleavage step, p43/p41 and p30 are processed to p10 and p20, respectively, which leads to the generation of the active caspase-8 heterotetramer (p20/p10)2.

Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Rat IgG1kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR