Anti-TIE-2 Monoclonal Antibody [clone: tek2]

Supplier: Adipogen
AG-20T-0102-C100
102978-600EA 636.92 USD
102978-600
Anti-TIE-2 Monoclonal Antibody [clone: tek2]
Antibodies
TIE-2 acts as cell-surface receptor for ANGPT1, ANGPT2 and ANGPT4 and regulates angiogenesis, endothelial cell survival, proliferation, migration, adhesion and cell spreading, reorganization of the actin cytoskeleton, but also maintenance of vascular quiescence. It has anti-inflammatory effects by preventing the leakage of proinflammatory plasma proteins and leukocytes from blood vessels. It is required for normal angiogenesis and heart development during embryogenesis and for post-natal hematopoiesis. After birth, it activates or inhibits angiogenesis, depending on the context. It inhibits angiogenesis and promotes vascular stability in quiescent vessels, where endothelial cells have tight contacts. In migrating endothelial cells that lack cell-cell adhesions, ANGT1 recruits TEK to contacts with the extracellular matrix, leading to the formation of focal adhesion complexes, activation of PTK2/FAK and of the downstream kinases MAPK1/ERK2 and MAPK3/ERK1 and ultimately to the stimulation of sprouting angiogenesis.

Type: Primary
Antigen: TIE-2
Clonality: Monoclonal
Clone: tek2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR