Anti-NOTCH1 Monoclonal Antibody (Biotin) [clone: No: (22E5)]

Supplier: Adipogen
AG-20B-0051B-C100
102971-576EA 387.69 USD
102971-576
Anti-NOTCH1 Monoclonal Antibody (Biotin) [clone: No: (22E5)]
Antibodies
Notch signaling pathway regulates many different cell fate decisions in both vertebrate and invertebrate species. There are 5 canonical Notch ligands in mammals: Jagged-1, Jagged-2, DLL1, DLL3 and DLL4. These can bind to the four Notch receptors Notch 1-4. It is important for pattern formation during development such as neurogenesis, angiogenesis or myogenesis and regulates T cell development and stem cell maintenance. Notch signaling is also involved in cellular processes through-out adulthood. Signaling via Notch occurs between neighbouring cells and both the receptor and its ligands are transmembrane proteins.

Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: 2200000
Conjugation: Biotin
Epitope:
Host:
Isotype: Rat IgG2akappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR