Anti-CASP1 Monoclonal Antibody (Biotin) [clone: Bally-1]

Supplier: Adipogen
AG-20B-0048B-C100
102971-568EA 609.23 USD
102971-568
Anti-CASP1 Monoclonal Antibody (Biotin) [clone: Bally-1]
Antibodies
Caspase-1 is the best-described inflammatory caspase. It processes the cytokines interleukin-1beta (IL-1beta) and IL-18 and induces pyroptotic cell death. Caspase-1 is activated by multiprotein complexes called Inflammasomes in response to numerous stimuli that are detected through distinct inflammasomes. NLRC4 responds to cytosolic flagellin, murine NLRP1b responds to anthrax lethal toxin, AIM2 responds to cytosolic DNA and NLRP3 responds to a variety of agonists including crystals.

Type: Primary
Antigen: CASP1
Clonality: Monoclonal
Clone: Bally-1
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR