Anti-LTBETAR Monoclonal Antibody [clone: 4H8 WH2]

Supplier: Adipogen
AG-20B-0008PF-C100
102971-558EA 360 USD
102971-558
Anti-LTBETAR Monoclonal Antibody [clone: 4H8 WH2]
Antibodies
The LTbetaR activates two different NF-kappaB pathways that lead to distinct patterns of gene induction, including selected chemokines and the cytokine BAFF, which is essential for the survival of mature B lymphocytes. LTbetaR activates the classical NF-kappaB (relA/p50) pathway, like the type 1 TNF receptor (TNFR1), that regulates proinflammatory genes, like the chemokine MIP1beta. However, LTbetaR, unlike TNFR1, also activates the processing of p100 to form RelB/p52 complexes, which activate genes involved in lymphoid organ formation and lymphocyte survival.

  • preservative free

Type: Primary
Antigen: LTBETAR
Clonality: Monoclonal
Clone: 4H8 WH2
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Rat IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR