Anti-IgG Goat Polyclonal Antibody (IRDye® 680LT)

Supplier: Li-Cor
925-68029
102971-222EA 150.12 USD
102971-222
Anti-IgG Goat Polyclonal Antibody (IRDye® 680LT)
Antibodies
The antibody was purified from antisera by affinity chromatography using antigens coupled to agarose beads. Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule rat IgG and with the light chains of other rat immunoglobulins. No reactivity was detected against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reactivity with mouse, bovine, horse, human, and rabbit serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot applications. Recommended for: Western blot, proein array, immunohistochemistry, microscopy, 2D gel detection, and tissue section imaging. NOT RECOMMENDED for: small animal imaging, In-Cell Wesern assays, or On-Cell Western assays.

Western blot: 1:20000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680LT
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR