Chameleon™ Duo Pre-stained Protein Ladder, LI-COR®
Supplier: Li-Cor
928-60000
102971-126EA 283.85 USD
102971-126
Chameleon™ Duo Pre-stained Protein Ladder, LI-COR®
Electrophoresis Reagents Molecular Weight Markers
These Pre-stained Protein Ladder provides multi-colored, pre-stained bands for visual inspection and two-color near-infrared detection
  • Easy visualization of gel migration and protein size
  • Multi-colored, pre-stained bands
  • Includes 11 protein bands
Use the ladder for easy visualization of gel migration and protein size, and to orient gel or membrane quickly.
Includes 11 protein bands from 8 to 260 kDa.