WesternSure™ Pen, LI-COR®
Supplier: Li-Cor
926-91000
102971-112EA 80.74 USD
102971-112
WesternSure™ Pen, LI-COR®
Pens Lab Markers
The WesternSure Pen is used to annotate visible protein ladders prior to chemiluminescent Western blot detection.
  • Optimized for detection using the C-DiGit® Blot Scanner
  • Optimized for detection using the Odyssey® Fc Imaging System
A unique marker is used which delivers an ink which emits light when incubated with commonly-used chemilumi­nescent substrates, including WesternSure PREMIUM
The WesternSure Pen was used to mark the blue protein standards for chemiluminescent Western blot detection. The blot was exposed to WesternSure PREMIUM chemiluminescent substrate and imaged on Odyssey Fc Imaging System.
The ink is faintly visible for easy identification of marked membranes.