Anti-IgM Goat Polyclonal Antibody (IRDye® 680RD)

Supplier: Li-Cor
925-68180
102971-038EA 150.12 USD
102971-038
Anti-IgM Goat Polyclonal Antibody (IRDye® 680RD)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using mouse IgM covalently linked to agarose. Based on ELISA, FLISA, and/or flow cytometry, this antibody reacts with the heavy chain of mouse IgM. This antibody was tested by Dot Blot and/or solid phase adsorbed for minimal cross-reactivity with mouse IgG1, IgG2a, IgG3, and IgA, pooled human sera and purified human paraproteins. The conjugate has been specifically tested and qualified for Western blot applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, and small animal imaging.

Western blot: 1:15000; In-Cell Western Assay: 1:800

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: IRDye 680RD
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR