Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)

Supplier: Li-Cor
925-32232
102971-002EA 150.12 USD
102971-002
Anti-IgG Goat Polyclonal Antibody (IRDye® 800CW)
Antibodies
Isolation of specific antibodies was accomplished by affinity chromatography using human IgG covalently linked to agarose beads. Based on ELISA, this antibody reacts with the heavy and light chains of human IgG. This antibody was tested by Dot Blot and/or solid-phase adsorbed for minimal cross-reactivity with bovine, horse, and mouse serum proteins, but may cross-react with immunoglobulins from other species. The conjugate has been specifically tested and qualified for Western blot applications. Highly Recommended for: Western blot, In-Cell Western Assay, On-Cell Western Assay, protein array, immunohistochemistry, microsciopy, 2D gel detection, tissue section imaging, virus titration assay, FRET-based assays, and small animal imaging.

Western blot: 1:5000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: IRDye 800CW
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR