Human Aortic Endothelial Cells, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0014
102970-954EA 1055.54 USD
102970-954
Human Aortic Endothelial Cells, Lifeline Cell Technology
Cells
Lifeline® Normal Human Aortic Endothelial Cells are quality tested and demonstrate optimal reduced-serum.
  • No exposure to antimicrobials or phenol red
  • May be cultured with or without VEGF
  • Optimal reduced-serum growth over at least 15 population doublings
Lifeline® Normal Human Aortic Endothelial Cells are quality tested in VascuLife® EnGS 2% serum medium and demonstrate optimal reduced-serum growth over a period of at least 15 population doublings at rates equal to or greater than other serum-supplemented media.
Lifeline® Aortic Endothelial Cells are not exposed to antimicrobials or phenol red when cultured in VascuLife® medium. Lifeline® Aortic Endothelial Cells may be cultured with or without VEGF.