Human Bladder Fibroblasts, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0050
102970-932EA 837.69 USD
102970-932
Human Bladder Fibroblasts, Lifeline Cell Technology
Cells
Lifeline® Normal Human Bladder Fibroblasts are quality tested and demonstrate optimal growth.
  • Tested for morphology
  • Can be grown in serum-free, phenol red-free, and antimicrobial-free conditions
  • Quality tested for optimal growth
Lifeline® normal Human Bladder Fibroblasts are quality tested in FibroLife® S2 or FibroLife® Xeno-Free Medium and demonstrate optimal growth at rates equal to or greater than classical serum-supplemented media
Bladder fibroblasts can be grown in serum-free, phenol red-free, and antimicrobial-free conditions when cultured in FibroLife® Serum-Free Medium.
Lifeline® bladder fibroblasts are tested for proper growth and morphology over a period of at least 15 population doublings.