AdipoLife™ DfKt™-2 Adipogenesis Kit (for Wharton's Jelly, Bone Marrow MSC), Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LL-0059
102970-904EA 250.62 USD
102970-904
AdipoLife™ DfKt™-2 Adipogenesis Kit (for Wharton's Jelly, Bone Marrow MSC), Lifeline Cell Technology
Cell Separation Media
AdipoLife™ DfKt™-2 Adipogenesis Medium is formulated to induce the adipogenic differentiation of Human Mesenchymal Stem Cells.
  • Induces the adipogenic differentiation of Human Mesenchymal Stem Cells
  • Derived from adipose tissue, bone marrow, and Wharton's Jelly
  • No antimicrobials
  • No phenol red
AdipoLife™ DfKt™-2 Adipogenesis Medium is a Lifeline® medium kit formulated to induce the adipogenic differentiation of Human Mesenchymal Stem Cells derived from adipose tissue, bone marrow, and Wharton's Jelly.
AdipoLife™ DfKt-2™ Adipogenesis Medium Complete Kit is offered in a kit format composed of AdipoLife™ Basal Medium (100mL) and a combined solution of differentiation factors called DifFactor™ 3. DifFactor™ 3 contains: rh Insulin, Indomethacin, 3-Isobutyl-1-Methylxanthine, Dexamethasone, Horse Serum (5% final conc.), L-Glutamine, and Ascorbic Acid.
AdipoLife™ DfKt™-2 contains no antimicrobials and no phenol red.
Ordering information: Use AdipoLife™ Adipogenesis Kit -2 with Wharton's Jelly and Bone Marrow Mesenchymal Stem Cells.