Bronchial/Tracheal Smooth Muscle Cells, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
FC-0059
102970-712EA 838 USD
102970-712
Bronchial/Tracheal Smooth Muscle Cells, Lifeline Cell Technology
Cells
Lifeline® Normal Human Bronchial/Tracheal Smooth Muscle Cells are quality tested and demonstrate optimal low serum growth.
  • Quality tested in VascuLife® SMC Medium
  • Demonstrate optimal low serum growth
  • Can be grown without antimicrobials or phenol red
Lifeline® Normal Human Bronchial/Tracheal Smooth Muscle Cells are quality tested in VascuLife® SMC Medium and demonstrate optimal low serum growth over a period of at least 15 population doublings at rates equal to or greater than other serum-supplemented media.
Bronchial Tracheal Smooth Muscle Cells can be grown without phenol red or antimicrobials when cultured in VascuLife® SMC Medium.