QuickTiter™ Lentivirus Titer Kit (Lentivirus-Associated p24 ELISA), Cell Biolabs
Supplier: Cell Biolabs
VPK-107-5 VPK-107-T VPK-107
102965-344EA 3661.54 USD
102965-344 102965-346 102970-592
QuickTiter™ Lentivirus Titer Kit (Lentivirus-Associated p24 ELISA), Cell Biolabs
Assays ELISAs
The QuickTiter™ Lentivirus Titer Kit (Lentivirus-Associated HIV p24) provides a unique and more accurate way to measure physical lentiviral titer
This kit substantially minimizes the problem of overestimating lentiviral titer with traditional p24 ELISA kits. The traditional p24 ELISA detects both virus-associated p24 and free p24 generated by 293 cells during transient transfection. Free p24 can account for a substantial portion of total p24 in the supernatant. Therefore, the ELISA typically overestimates the quantity of lentivirus present