Ultrafree-MC GV Centrifugal Filter, MilliporeSigma
Supplier: MilliporeSigma
UFC30GV0S
102970-584PK 364.62 USD
102970-584
Ultrafree-MC GV Centrifugal Filter, MilliporeSigma
Filters Centrifugal Filters
Ultrafree® centrifugal devices are single-use, disposable filters used for removing particulates from aqueous biological solutions.
  • Five different pore sizes from 0.1 to 5.0 µm
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 1.5 ml tubes
These filters are used for the removal of particles and precipitates from aqueous and some solvent-based samples. Ideal for use with protein and nucleic acid solutions.