Anti-Fx1A Rabbit Polyclonal Antibody

Supplier: Bioss

BS-1338R
10297-362EA 659.44 USD
10297-362
Anti-Fx1A Rabbit Polyclonal Antibody
Antibodies
Binding of anti-Fx1A to Heymann nephritis antigens (HA) on rat glomerular epithelial cells (GECs) in culture leads to capping and disappearance of antigens from the cell surface. This process may contribute to the formation of glomerular subepithelial immune deposits in vivo. The authors differentially extracted GECs to determine whether HA redistribution is mediated by cytoskeletal components. Observations were made by phase-contrast and immunofluorescence microscopy on primary and passaged GECs in monolayer culture and by spectrofluorimetry on GECs in suspension.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Fx1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR