a1-3,4 Fucosidase, New England Biolabs
Supplier: New England Biolabs (NEB)
P0769S
102969-118EA 157 USD
102969-118
a1-3,4 Fucosidase, New England Biolabs
Protein Purification Protein Purification Reagents
α1-3,4 Fucosidase, (also known as AMF) is a broad specificity exoglycosidase that catalyzes the hydrolysis of α1-3 and α1-4 linked fucose residues from oligosaccharides and glycoproteins.