Total Carbohydrate Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-682
102966-256EA 523.08 USD
102966-256
Total Carbohydrate Assay Kit, Cell Biolabs
Assays Cellular Assays
Cell Biolabs' Total Carbohydrate Assay detects total carbohydrates within samples based on the phenol-sulfuric acid method
Carbohydrates are hydrolyzed to furfural and furfural derivates upon addition of sulfuric acid, which then react with the Developing Solution to form a chromagen that is detected at 490nm. Total carbohydrate levels in an unknown sample are calculated based on a glucose standard curve.