Hydroxyproline Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-675
102966-250EA 572.31 USD
102966-250
Hydroxyproline Assay Kit, Cell Biolabs
Assays Cellular Assays
Cell Biolabs' Hydroxyproline Assay Kit is a quantitative colorimetric assay for measuring the hydroxyproline concentration in protein samples.  Hydroxyproline in acid hydrolyzed samples is converted to pyrrole through a reaction with Chlormine T, which then reacts with Ehrlich’s Reagent to form a chromophore that is detected at 540-560nm
Hydroxyproline levels in samples are calculated based on a hydroxyproline standard curve. Detect hydroxyproline levels as low as 50 µM.