Glutamate Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-674
102966-248EA 572.31 USD
102966-248
Glutamate Assay Kit, Cell Biolabs
Assays Enzyme Assays
Cell Biolabs' Glutamate Assay Kit is a quantitative, fluorometric assay that uses glutamate specific enzymes to generate hydrogen peroxide
A fluorometric probe is oxidized by hydrogen peroxide to generate fluorescent Resorufin, which correlates to the level of glutamate in the sample. Glutamate levels in an unknown sample are calculated based on a glutamate standard curve.