Anti-Carbamyl lysine Goat Polyclonal Antibody

Supplier: Cell Biolabs
STA-077
102966-226EA 381.54 USD
102966-226
Anti-Carbamyl lysine Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Carbamyl lysine
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR