pMZs Retroviral Expression Vector, Cell Biolabs
Supplier: Cell Biolabs
RTV-030
102966-176EA
652.31
USD
102966-176
pMZs Retroviral Expression Vector, Cell Biolabs
Vectors, Plasmids and Libraries
Cell Biolabs' pMZs vector is suitable for making retrovirus for general gene expression
Clone your gene of interest into one of these vectors and then transfect the construct into a retroviral packaging cell line, 293T cells, or 293RTV cells.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)