OxiSelect™ HORAC Activity Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
The Hydroxyl Radical Antioxidant Capacity (HORAC) assay is a powerful tool to measure the antioxidant capacity of biomolecules
OxiSelect™ HORAC Activity Assay measures this capacity in less than 2 hours from a wide variety of sample types. The assay is suitable for use with plasma, cell fractions, and tissue lysates, as well as solid and aqueous nutrition samples. Obtain results in about 2 hours.