293RTV Cell Line, Cell Biolabs
Supplier: Cell Biolabs
RV-100
102965-754EA 523.08 USD
102965-754
293RTV Cell Line, Cell Biolabs
Cells
The 293RTV Cell Line is derived from the parental 293T cells but selected for attributes that increase retroviral production, including firmer attachment and larger surface area
This allows the cells to produce higher levels of lentivirus compared to standard 293T cells.