AAV-3 Helper Free Expression Complete Systems, Cell Biolabs
Supplier: Cell Biolabs
VPK-411-SER3 VPK-415-SER3 VPK-416-SER3 VPK-417-SER3 VPK-418-SER3 VPK-419-SER3 VPK-430-SER3 VPK-410-SER3
102965-474EA 1347.69 USD
102965-474 102965-492 102965-506 102965-520 102965-534 102965-548 102965-566 102966-354
AAV-3 Helper Free Expression Complete Systems, Cell Biolabs
Vectors, Plasmids and Libraries
AAV-3 Helper Free Expression Complete Systems contain the following constructs needed to package your recombinant serotype-3 adeno-associated virus: an expression vector, pHelper plasmid, a serotype-specific Rep-Cap plasmid, and an optional GFP expression vector for making a control virus
AAV Helper Free Expression Complete Systems are available for native serotypes 1 through 6. The shRNA expression vectors contain a U6 promoter. All other expression vectors contain a CMV promoter which is optimal for general gene expression, with the exception of our promoterless vector which is useful if you wish to clone in a gene-specific promoter along with your gene of interest.