OxiSelect™ Protein Carbamylation Sandwich ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-877
102965-328EA 910.77 USD
102965-328
OxiSelect™ Protein Carbamylation Sandwich ELISA Kit, Cell Biolabs
Assays ELISAs
The OxiSelect™ Protein Carbamylation Sandwich ELISA Kitis an enzyme immunoassay developed for rapid detection and quantitation of protein carbamylation
CBL-BSA standards or protein samples are added to an Anti-CBL Antibody Coated Plate for 2 hours at 37ºC.  The CBL protein adducts present in the sample or standard are probed with an anti-CBL antibody, followed by an HRP conjugated secondary antibody.  The protein CBL adduct content in an unknown sample is determined by comparing against a standard curve prepared from CBL-BSA standards. This kit can detect as little as 1.5 ng/mL of CBL-BSA.