OxiSelect™ Nε-(carboxyethyl) lysine (CEL) Competitive ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-813
102965-282EA 929.23 USD
102965-282
OxiSelect™ Nε-(carboxyethyl) lysine (CEL) Competitive ELISA Kit, Cell Biolabs
Assays ELISAs
OxiSelect™ Nε-(carboxyethyl) lysine (CEL) ELISA Kit provides rapid detection and quantitation of CEL protein adducts
First, a CEL conjugate is coated on the ELISA plate.  The unknown CEL protein samples or CEL-BSA standards are then added to the CEL conjugate preabsorbed plate.  After a brief incubation, the anti-CEL monoclonal antibody is added, followed by an HRP conjugated secondary antibody.  The content of CEL protein adducts in unknown samples is determined by comparison with the predetermined CEL-BSA standard curve.  Despite the structure similarity between CEL and CML, the anti-CEL specific antibody in the OxiSelect™ CEL ELISA Kit will not cross react with CML protein adducts.