OxiSelect™ Myeloperoxidase Activity Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-803
102965-268EA 609.23 USD
102965-268 102965-272
OxiSelect™ Myeloperoxidase Activity Assay Kits, Cell Biolabs
Assays Cellular Assays
OxiSelect™ Myeloperoxidase Activity Assay Kits are quantitative assays for measuring the myeloperoxidase activity within a sample
OxiSelect™ Myeloperoxidase Chlorination Activity Assay Kits are quantitative assays for measuring the chlorination activity of myeloperoxidase within a sample.  Kits are available with either colorimetric or fluorometric detection. The MPO enzyme catalyzes the reaction of hydrogen peroxide (H2O2) with chloride ions to create hypochlorous acid (HOCl), which results in a reaction with the included probe to produce a color change or fluorescence readout.
The OxiSelect™ Myeloperoxidase Peroxidation Activity Assay Kit is a quantitative, fluorometric assay for measuring the myeloperoxidase peroxidation activity within a sample.  In the presence of MPO, hydrogen peroxide (H2O2) oxidizes the non-fluorescent probe to a highly fluorescent form which is detected in a 96-well fluorescence plate reader.
Myeloperoxidase has both chlorination and peroxidation activity, and these activities are measured separately.