Phosphatidylcholine Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-600
102965-204EA
578.46
USD
102965-204
Phosphatidylcholine Assay Kit, Cell Biolabs
Assays
Enzyme Assays
Cell Biolabs’ Phosphatidylcholine Assay Kit measures the phosphatidylcholine present within serum, plasma, or tissue samples
Samples are compared to a known concentration of phosphatidylcholine standard within the 96-well microtiter plate format. Samples and standards are incubated for 60 minutes and then read with a standard 96-well fluorometric plate reader. Detect phosphatidylcholine concentrations as low as 0.5 mg/dL.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)