Cdc42 Activation Assays, Cell Biolabs
Supplier: Cell Biolabs
STA-402
102965-110EA 855.38 USD
102965-110
Cdc42 Activation Assays, Cell Biolabs
Assays Protein Assays
Cell Biolabs' Cdc42 Activation Assays use visible agarose beads to selectively precipitate the active form of Cdc42 protein
The precipitated small GTPase is then detected by Western blot using a Cdc42-specific antibody included in the kit. The assay uses a safe, non-radioactive format.