Rac Activation Assays, Cell Biolabs
Supplier: Cell Biolabs
STA-401-2
102965-102EA 640 USD
102965-102 102965-104 102965-106
Rac Activation Assays, Cell Biolabs
Assays Protein Assays
Cell Biolabs' Rac Activation Assays use visible agarose beads to selectively precipitate the active form of Rac
The precipitated small GTPase is then detected by Western blot using a Rac1- or Rac2-specific antibody included in the kit. The assays use a safe, non-radioactive format.