Uric Acid/Uricase Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-375
102965-042EA 726.15 USD
102965-042
Uric Acid/Uricase Assay Kit, Cell Biolabs
Assays Immunological Assays
Cell Biolabs’ Uric Acid/Uricase Assay Kit is a simple HTS-compatible assay for measuring uric acid concentrations in biological samples such as serum, plasma, and urine without any need for pretreatment.  The kit has a detection sensitivity limit of 0.5 µM of uric acid or 1 mU/mL of uricase
 Each kit provides sufficient reagents to perform up to 400 assays, including standard curve and unknown samples.