OxiSelect™ Comet Assay Kits and Slides (3-Well), Cell Biolabs
Supplier: Cell Biolabs
OxiSelect™ 3-Well Comet Assay Kits and Slides provide a fast, convenient way to screen for general DNA damage, regardless of the source or nature of the damage
Kits include Comet Slides, reagents, and a fluorescent dye to visualize cells under an epifluorescence microscope. Slides are available to order separately.