OxiSelect™ UV-Induced DNA Damage ELISA Kit (6-4PP Quantitation), Cell Biolabs
Supplier: Cell Biolabs
OxiSelect™ UV-Induced DNA Damage ELISA Kit measures 6-4PP formation in DNA isolated from cells or tissue
Standards or unknown DNA samples are first heat denatured before adsorbed onto a 96-well DNA binding plate. The sample or standard is then probed with an anti-6-4PP antibody, followed by an HRP conjugated secondary antibody.