Anti-N-epsilon-(carboxymethyl) lysine Goat Polyclonal Antibody

Supplier: Cell Biolabs
STA-013
102964-792EA 486.15 USD
102964-792
Anti-N-epsilon-(carboxymethyl) lysine Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: N-epsilon-(carboxymethyl) lysine
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR