ViraDuctin™ Retrovirus Transduction Kit, Cell Biolabs
Supplier: Cell Biolabs
Cell Biolabs' ViraDuctin™ Retrovirus Transduction Kit provides superior retroviral transduction efficiencies in a variety of cell lines, even when compared to transductions in the presence of Polybrene®
This system is ideal for many primary cells as well as immobilized cells. The kit uses a proprietary reagent cocktail that forms supercomplex with retrovirus. The result is an increase in infection efficiency of 2- to 6-fold. The kit is espeically useful for transduction of non-permissive cells, including many primary cells and stem cells.