pBABE Retroviral Expression Vectors, Cell Biolabs
Supplier: Cell Biolabs
RTV-001-HYGRO
102964-494EA 547.69 USD
102964-494 102964-498 102964-502 102964-504
pBABE Retroviral Expression Vectors, Cell Biolabs
Vectors, Plasmids and Libraries
Cell Biolabs' pBABE vectors are suitable for making retrovirus for general gene expression in various cells
Clone your gene of interest into one of these vectors and then transfect the construct into a retroviral packaging cell line, 293T cells, or 293RTV cells.