StemTAG™ Alkaline Phosphatase Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
CBA-300 CBA-301 CBA-302
102963-864EA 486.15 USD
102963-864 102963-866 102963-868 102966-504 102966-506 102966-508
StemTAG™ Alkaline Phosphatase Assay Kits, Cell Biolabs
Assays Enzyme Assays
The StemTAG™ Alkaline Phosphatase Assay Kits provide reagents for monitoring alkaline phosphatase activity via either immunocytochemistry staining or 96-well plate quantitation with either colorimetric or fluorescence readout
For best value choose one of our Alkaline Phosphatase Combo Kits that provide reagents for both ICC and 96-well plate detection.