Quantitative Cellular Senescence Assay, Cell Biolabs
Supplier: Cell Biolabs
Cell Biolabs' Cellular Senescence Flow Cytometry Assay provides an efficient method to measure Senescence Associated (SA) ß-galactosidase activity
A fluorogenic substrate is added directly to senescent cells in a standard 35 mm culture dish. Results can be measured by either flow cytometry or epifluorescence microscope.