CytoSelect™ Cell Invasion Assay Kits, Collagen Coated, Cell Biolabs
Supplier: Cell Biolabs
CBA-110-COL CBA-111-COL CBA-112-COL
102963-748EA 689.23 USD
102963-748 102963-756 102963-762
CytoSelect™ Cell Invasion Assay Kits, Collagen Coated, Cell Biolabs
Assays Cellular Assays
CytoSelect™ Cell Invasion Assays are ideal for determining the invasive properties of cells
With the CytoSelect™ Cell Invasion Assays, no manual cell counting is required. Quantitation of cells is performed using a colorimetric or fluorometric dye in a 96-well plate reader. The 8 µm pore size is suitable for most cell types including epithelial cells, fibroblasts, and cancer cell lines. Kits are available in 24-well and 96-well formats.
These kits use a Boyden Chamber in which inserts containing a membrane with a defined pore size are nested in the wells of a cell culture plate. The top of the insert membrane is coated with a thin layer of collagen gel. Invasive cells are seeded above the membrane and must degrade the collagen layer before migrating toward a chemoattractant in the lower chamber.