Anti-GFP Mouse Monoclonal Antibody [clone: GF28R]

Supplier: Cell Biolabs
AKR-020
102963-602EA 498.46 USD
102963-602
Anti-GFP Mouse Monoclonal Antibody [clone: GF28R]
Antibodies
Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: GF28R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Aequorea victoria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR