Anti-V5 Mouse Monoclonal Antibody [clone: E10]

Supplier: Cell Biolabs
AKR-008
102963-598EA 486.15 USD
102963-598
Anti-V5 Mouse Monoclonal Antibody [clone: E10]
Antibodies
Type: Primary
Antigen: V5
Clonality: Monoclonal
Clone: E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR