Anti-GST Mouse Monoclonal Antibody [clone: GST.B6]

Supplier: Cell Biolabs
AKR-005
102963-592EA 486.15 USD
102963-592
Anti-GST Mouse Monoclonal Antibody [clone: GST.B6]
Antibodies
Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone: GST.B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR